missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human TCEB2 Protein

Product Code. 18214805 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214805 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214805 Supplier Novus Biologicals™ Supplier No. NBC1225790.5MG
  • ONLINE EXCLUSIVE PRICE
    1160.00 EUR
    1096.20 EUR / 0.5mg
    Save 63.80 EUR
    5% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-118 of Human TCEB2 The Recombinant Human TCEB2 Protein is derived from E. coli. The Recombinant Human TCEB2 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_009039
Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 1mMDTT, 0.1mM PMSF, 20% glycerol
Gene ID (Entrez) 6923
Molecular Weight (g/mol) 13.1kDa
Purification Method Protein
Quantity 0.5 mg
Source Recombinant TCEB2 was expressed as insoluble protein aggregate in E. coli
Immunogen Elongin B, 1-118aa, Human, Recombinant, E. coli (NP_009039).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name TCEB2
Conjugate Unconjugated
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.