Learn More
Novus Biologicals™ HEK293 Cell Lysate
Product Code. 18004314 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18004314 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18004314 Supplier Novus Biologicals™ Supplier No. NBP225048
Applications: SDS-Page
The HEK293 Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mg
Storage Buffer 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol
Test Specificity This lysate was prepared from a cell culture of HEK 293 (human embryonic kidney cells).
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Research Use Only