missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Annexin A8/ANXA8 Protein

Product Code. 18214853 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214853 0.5 mg 0.5mg
18225622 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18214853 Supplier Novus Biologicals™ Supplier No. NBP1483280.5MG
  • ONLINE EXCLUSIVE PRICE
    1090.00 EUR
    1027.95 EUR / 0.5mg
    Save 62.05 EUR
    5% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-327 of Human Annexin A8/ANXA8 The Recombinant Human Annexin A8/ANXA8 Protein is derived from E. coli. The Recombinant Human Annexin A8/ANXA8 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001035173
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 1mM DTT, 1mM EDTA, 10% glycerol
Gene ID (Entrez) 653145
Molecular Weight (g/mol) 39.0kDa
Purification Method Protein
Quantity 0.5 mg
Source Recombinant human ANXA8 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen ANXA8, 1-327aa, Human, His tag, E. coli (NP_001035173).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Annexin A8/ANXA8
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.