missing translation for 'onlineSavingsMsg'
Learn More

Nifluridide, TRC

Product Code. 30464996
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30464996 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464996 Supplier Toronto Research Chemicals Supplier No. TRCN43158010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61444-62-0
Molecular Formula C10 H6 F7 N3 O3
Molecular Weight (g/mol) 349.16
Synonym N-[2-Amino-3-nitro-5-(trifluoromethyl)phenyl]-2,2,3,3-tetrafluoropropanamide, Compound 109168, EL 468, EL 968, L 27, L 27 (pesticide), N1-(2,2,3,3-Tetrafluoropionyl)-3-nitro-5-trifluoromethyl-o-phenylenediamine, N1-(2,2,3,3-Tetrafluoropropionyl)-3'-nitro-5'-trifluoromethyl-o-phenylenediamine, Nifluridide
IUPAC Name N-[2-amino-3-nitro-5-(trifluoromethyl)phenyl]-2,2,3,3-tetrafluoropropanamide
SMILES Nc1c(NC(=O)C(F)(F)C(F)F)cc(cc1[N+](=O)[O-])C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H6F7N3O3/c11-7(12)9(13,14)8(21)19-4-1-3(10(15,16)17)2-5(6(4)18)20(22)23/h1-2,7H,18H2,(H,19,21)
Formula Weight 349.0297
Chemical Name or Material Nifluridide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.