CAS RN 61444-62-0
CAS RN 61444-62-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nifluridide, TRC
CAS: 61444-62-0 Molecular Formula: C10 H6 F7 N3 O3 Molecular Weight (g/mol): 349.16 Synonym: N-[2-Amino-3-nitro-5-(trifluoromethyl)phenyl]-2,2,3,3-tetrafluoropropanamide,Compound 109168,EL 468,EL 968,L 27,L 27 (pesticide),N1-(2,2,3,3-Tetrafluoropionyl)-3-nitro-5-trifluoromethyl-o-phenylenediamine,N1-(2,2,3,3-Tetrafluoropropionyl)-3'-nitro-5'-trifluoromethyl-o-phenylenediamine,Nifluridide IUPAC Name: N-[2-amino-3-nitro-5-(trifluoromethyl)phenyl]-2,2,3,3-tetrafluoropropanamide SMILES: Nc1c(NC(=O)C(F)(F)C(F)F)cc(cc1[N+](=O)[O-])C(F)(F)F