missing translation for 'onlineSavingsMsg'
Learn More

(-)-Nicotine Tartrate (White Solid), Fisher BioReagents

Product Code. p-171916 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10717935 250 mg Amber Glass 250mg
10566764 1 g Amber Glass 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10717935 Supplier Fisher BioReagents Supplier No. BP2533250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
(-)-Nicotine Tartrate is used in biochemical research applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-31-6
Molecular Formula C18H26N2O12
Molecular Weight (g/mol) 462.408
InChI Key RFEJUZJILGIRHQ-USHJUENDSA-N
Synonym nicotine ditartrate
PubChem CID 122176683
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid;(3S)-2,3-dihydroxybutanedioic acid;3-(1-methylpyrrolidin-2-yl)pyridine
SMILES CN1CCCC1C2=CN=CC=C2.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 98.00°C
Color White
Packaging Amber Glass
Quantity 250 mg
Merck Index 15, 6609
Specific Rotation Report ° (+ or -)
Formula Weight 462.4
Physical Form Solid
Chemical Name or Material (-)-Nicotine Tartrate
compliance-icons compliance-icons
Product Identifier
  • (-)-Nicotine Tartrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.