accessibility menu, dialog, popup

UserName

CH3NNOH(R)(R)OHHOOOHOOH(R)(R)OHHOOOHO

CAS RN 65-31-6

IUPAC Name: bis((2R,3R)-2,3-dihydroxybutanedioic acid); 3-(1-methylpyrrolidin-2-yl)pyridine
Synonyms: bis(L(+)-tartaric acid); nicotine
Molecular Weight (g/mol): 462.41
Molecular Formula: C18H26N2O12
InChi Key: RFEJUZJILGIRHQ-WBPXWQEISA-N
SMILES: O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCCC1C1=CC=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results