missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester, 95%

Product Code. p-7055002
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448326 1 g 1g
15458326 5 g 5g
15468326 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448326 Supplier Thermo Scientific Alfa Aesar Supplier No. H63470.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 34404-36-9 | C23H31N3O8 | 477.51 g/mol

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester is a reactant involved in Cellular labeling and tumor MRI as a bimodal lipidic contrast agent, Synthesis of improved peptide prodrugs 5-ALA for photodynamic therapy, Design of high affinity peptide conjugates as markers for small peptide transporter PEPT1 (SLC15A1), Preparation of human growth hormone derivatives containing PEG groups, Enantiodivergent asymmetric cyclization and Solid-phase synthesis of peptide-cationic steroid conjugates for antibacterial screening.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34404-36-9
Molecular Formula C23H31N3O8
Molecular Weight (g/mol) 477.51
MDL Number MFCD00037915
InChI Key YWLICOCXPNQJPC-AGDOHHJYNA-N
Synonym boc-lys z-osu, s-2,5-dioxopyrrolidin-1-yl 6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys cbz-osu, 2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys z osu, boc-lys z-o su, boc-lys-z-osu, n,a-boc-n, a-z-l-lysine hydroxysuccinimide ester, n,a-z-n, a-boc-l-lysine hydroxysuccinimide ester, a-cbz-l-lysine n-hydroxysuccinimide ester
PubChem CID 12686578
IUPAC Name 2,5-dioxopyrrolidin-1-yl 6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoate
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 108°C to 117°C
Color White
Beilstein 1559393
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 477.51
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.