accessibility menu, dialog, popup

UserName

CH3CH3H3COONH(S)NHOOOONOO

CAS RN 34404-36-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 6-{[(benzyloxy)carbonyl]amino}-2-[(tert-butoxycarbonyl)amino]hexanoate
Molecular Weight (g/mol): 477.51
Molecular Formula: C23H31N3O8
InChi Key: YWLICOCXPNQJPC-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results