missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NDM-1 Polyclonal specifically detects NDM-1 in Bacteria, E. coli, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | NDM-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873] |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mL |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?