missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NCF4 Polyclonal antibody specifically detects NCF4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | NCF4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | MGC3810, NCF, NCF-4, neutrophil cytosol factor 4, neutrophil cytosolic factor 4 (40kD), neutrophil cytosolic factor 4, 40kDa, Neutrophil NADPH oxidase factor 4, p40phox, p40-phox, SH3 and PX domain-containing protein 4, SH3PXD4P40PHOX |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-190 of human NCF4 (NP_038202.2).,, Sequence:, EIAEMRIPALNAYMKSLLSLPVWVLMDEDVRIFFYQSPYDSEQVPQALRRLRPRTRKVKSVSPQGNSVDRMAAPRAEALFDFTGNSKLELNFKAGDVIFLL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?