Learn More
NAP 226-90, TRC
Product Code. 30136320
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136320 1 g 1g
30135444 100 mg 100mg
17943905 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136320 Supplier Toronto Research Chemicals Supplier No. TRCN3250001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 139306-10-8
Molecular Formula C10 H15 N O
Molecular Weight (g/mol) 165.23
Synonym Phenol, 3-[(1S)-1-(dimethylamino)ethyl]-, Phenol, 3-[1-(dimethylamino)ethyl]-, (S)-, (S)-3-[1-(Dimethylamino)ethyl]phenol, Rivastigmine Related Compound C (USP), NAP 226-90, 3-[(1S)-1-(Dimethylamino)ethyl]phenol, Dimetol
IUPAC Name 3-[(1S)-1-(dimethylamino)ethyl]phenol
SMILES C[C@H](N(C)C)c1cccc(O)c1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H15NO/c1-8(11(2)3)9-5-4-6-10(12)7-9/h4-8,12H,1-3H3/t8-/m0/s1
Formula Weight 165.1154
Chemical Name or Material Dimetol