accessibility menu, dialog, popup

UserName

CAS RN 139306-10-8

H3COHNCH3CH3

CAS RN 139306-10-8

IUPAC Name: 3-[1-(dimethylamino)ethyl]phenol
Synonyms: 3-[1-(dimethylamino)ethyl]phenol
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: GQZXRLWUYONVCP-UHFFFAOYNA-N
SMILES: CC(N(C)C)C1=CC(O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results