missing translation for 'onlineSavingsMsg'
Learn More

NAALADase-like 1/NAALADL1 Antibody (005), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18215059 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215059 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215059 Supplier Novus Biologicals Supplier No. NBP290747AF594
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NAALADase-like 1/NAALADL1 Monoclonal specifically detects NAALADase-like 1/NAALADL1 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAALADase-like 1/NAALADL1
Applications ELISA
Classification Monoclonal
Clone 5
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias I100, NAALADASEL, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse NAALADase-like 1/NAALADL1 (Q7M758; Pro29-Leu745).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10004
Test Specificity Has cross-reactivity in ELISA with: Human NAALADL1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.