missing translation for 'onlineSavingsMsg'
Learn More

N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxycytidine, 98%, Thermo Scientific™

Product Code. 15460377
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460377 1 g 1g
15470377 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460377 Supplier Thermo Scientific Alfa Aesar Supplier No. J64614.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1241724-98-0
Molecular Formula C37H34FN3O7
Molecular Weight (g/mol) 651.69
MDL Number MFCD15145192
InChI Key RAIBEZUVTIPFOJ-PFDTVBFTNA-N
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Color White to Yellow
Quantity 1 g
IUPAC Name N-[1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-3-fluoro-4-hydroxyoxolan-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl]benzamide
Formula Weight 651.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxycytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.