CAS RN 1241724-98-0
IUPAC Name:
N-[1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-3-fluoro-4-hydroxyoxolan-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl]benzamide
Synonyms:
N-[1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-3-fluoro-4-hydroxyoxolan-2-yl)-2-oxopyrimidin-4-yl]benzamide
Molecular Weight (g/mol):
651.69
Molecular Formula:
C37H34FN3O7
InChi Key:
RAIBEZUVTIPFOJ-UHFFFAOYNA-N
SMILES:
COC1=CC=C(C=C1)C(OCC1OC(C(F)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
1
–
1
of
1
results
1
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxycytidine, 98%, Thermo Scientific™
CAS: 1241724-98-0 Molecular Formula: C37H34FN3O7 Molecular Weight (g/mol): 651.69 MDL Number: MFCD15145192 InChI Key: RAIBEZUVTIPFOJ-PFDTVBFTNA-N SMILES: COC1=CC=C(C=C1)C(OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1