Learn More
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxycytidine, 98%, Thermo Scientific™
Product Code. 15470377
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470377 100 mg 100mg
15460377 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15470377 Supplier Thermo Scientific Alfa Aesar Supplier No. J64614.MC
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1241724-98-0
Molecular Formula C37H34FN3O7
Molecular Weight (g/mol) 651.69
MDL Number MFCD15145192
InChI Key RAIBEZUVTIPFOJ-PFDTVBFTNA-N
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
Specifications
Color White to Yellow
Quantity 100 mg
IUPAC Name N-{1-[(2R,3R,4R,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-3-fluoro-4-hydroxyoxolan-2-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
Formula Weight 651.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxycytidine