missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate, 98+%

Product Code. p-7036350
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431416 25 mg 25mg
15441416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15431416 Supplier Thermo Scientific Alfa Aesar Supplier No. H54627.MB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 64987-85-5 | C16H18N2O6 | 334.328 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64987-85-5
Molecular Formula C16H18N2O6
Molecular Weight (g/mol) 334.328
MDL Number MFCD00009634
InChI Key JJAHTWIKCUJRDK-UHFFFAOYSA-N
Synonym smcc, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate, trans-4-maleimidomethyl cyclohexanecarboxylic acid-nhs, unii-b357p1g1if, n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate, n-4-carboxycyclohexylmethyl maleimide n-hydroxysuccinimide ester, trans-2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-hydroxysuccinimidyl 4-n-maleimidomethylcyclohexane-1-carboxylate, succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate
PubChem CID 125175
ChEBI CHEBI:63174
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
SMILES C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O

Specifications

Melting Point 180°C to 182°C
Quantity 25 mg
Beilstein 1555271
Sensitivity Light and moisture sensitive
Formula Weight 334.33
Percent Purity ≥98%
Chemical Name or Material N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.