missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-β-D-Glucopyranoside, MP Biomedicals™

Product Code. 11406462 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Unit Size:
1g
250mg
500mg
5g
This item is not returnable. View return policy

Product Code. 11406462

Brand: MP Biomedicals 02150054.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

n-Octyl-β-D-Glucopyranoside >98%, white powder, non-ionic detergent for membrane protein.

  • Non-ionic alkyl-glucoside detergent commonly used in the solubilization of membrane proteins and phospholipids.
  • Used at a molar dependant ratios for the solubilization of phosphatidylcholine bilayers.
  • Used for protein crystallization, as well as for enhancement of oligosaccharide cleavage from glycoproteins using endoglycosidase F.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
Synonym n-Octyl-b-D-glucoside, OGP
PubChem CID 62852
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Physical Form Powder
Melting Point 89°C to 100°C (literature)
Quantity 250 mg
Specific Rotation -31 ± 3° (c=5, water)
Beilstein 84118EC
Solubility Information Soluble in water (10mg/mL-clear colorless solution).
Grade 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.