accessibility menu, dialog, popup

UserName

H3CO(R)O(R)OH(S)OH(S)OH(R)HO

CAS RN 29836-26-8

IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(octyloxy)oxane-3,4,5-triol
Synonyms: octyl glucoside caprylyl glucoside
Molecular Weight (g/mol): 292.37
Molecular Formula: C14H28O6
InChi Key: HEGSGKPQLMEBJL-RKQHYHRCSA-N
SMILES: CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
Quantity 
  • (6)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (5)
  • (4)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)
  • (3)