missing translation for 'onlineSavingsMsg'
Learn More

n-Nonadecane, Sigma-Aldrich™

Product Code. 15841671
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15841671 25 g 25g
15806710 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15841671 Supplier Sigma Aldrich Supplier No. 8221100025
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-92-5

Specifications

Melting Point 31°C to 34°C
Density 0.79 g/cm3 at 20°C
Boiling Point 329°C to 331°C
Molecular Formula CH3(CH2)17CH3
MDL Number MFCD00009012
Quantity 25 g
Synonym n-Nonadecane, n-Nonadecane
Molecular Weight (g/mol) 268.52
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.