accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 629-92-5

IUPAC Name: nonadecane
Synonyms: nonadecane nonadekan
Molecular Weight (g/mol): 268.53
Molecular Formula: C19H40
InChi Key: LQERIDTXQFOHKA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)