missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Dimethyl Acetal, Sigma-Aldrich™

Product Code. 15828920
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15828920 100 mL 100mL
15818920 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15828920 Supplier Sigma Aldrich Supplier No. 8030680100
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

N, N-Dimethylformamide dimethyl acetal (DMF-DMA) plays an important role in organic synthesis due to its higher reactivity. As a methylating agent, DMF-DMA has been used in the synthesis of methyl esters from acids, methyl ethers, and thioethers from phenols and aromatic thiols. DMF-DMA has been used as a formulating agent to synthesize enamines from active methylenes and active methyl groups, and amidines from amines and amides or thioamide. It is also possible to produce heterocyclic compounds using DMF-DMA by cyclizing two functional groups.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4637-24-5

Specifications

Melting Point -85°C
Density 0.895 g/cm3 at 20°C
Molecular Formula (CH3O)2CHN(CH3)2
MDL Number MFCD00008482
Quantity 100 mL
Synonym N,N-Dimethylformamide dimethyl acetal, N,N-Dimethyldimethoxymethylamine, DMF-DMA, N,N-Dimethylformamide dimethyl acetal
Molecular Weight (g/mol) 119.16
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.