accessibility menu, dialog, popup

UserName

CAS RN 4637-24-5

H3COOCH3NCH3H3C

CAS RN 4637-24-5

IUPAC Name: (dimethoxymethyl)dimethylazanium
Synonyms: (dimethoxymethyl)dimethylazanium
Molecular Weight (g/mol): 120.17
Molecular Formula: C5H14NO2
InChi Key: ZSXGLVDWWRXATF-UHFFFAOYSA-O
SMILES: COC(OC)[NH+](C)C
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)