missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di-n-hexylacetamide, 98+%, Thermo Scientific™

Product Code. 11333006
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333006 10 g 10g
11343006 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333006 Supplier Thermo Scientific Alfa Aesar Supplier No. L10805.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16423-51-1
Molecular Formula C14H29NO
Molecular Weight (g/mol) 227.39
MDL Number MFCD00082658
InChI Key JFUCOPIALLYRGN-UHFFFAOYSA-N
Synonym n,n-di-n-hexylacetamide, n-dihexyl-acetamid, acetamide, n,n-dihexyl, n,n-di-n-hexylacetamide, 98+%
PubChem CID 243972
IUPAC Name N,N-dihexylacetamide
SMILES CCCCCCN(CCCCCC)C(C)=O

Specifications

Density 0.86
Boiling Point 130°C (2 mmHg)
Flash Point 106°C (223°F)
Refractive Index 1.451
Quantity 10 g
Beilstein 1772656
Formula Weight 227.39
Percent Purity ≥98%
Chemical Name or Material N,N-Di-n-hexylacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.