accessibility menu, dialog, popup

UserName

CAS RN 16423-51-1

H3CNCH3CH3O

CAS RN 16423-51-1

IUPAC Name: N,N-dihexylacetamide
Synonyms: N,N-dihexylacetamide
Molecular Weight (g/mol): 227.39
Molecular Formula: C14H29NO
InChi Key: JFUCOPIALLYRGN-UHFFFAOYSA-N
SMILES: CCCCCCN(CCCCCC)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results