missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(1,3,4-thiadiazol-2-yl)methanediamine, TRC

Product Code. 18091520
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18091520 1 g 1g
17979839 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18091520 Supplier Toronto Research Chemicals Supplier No. TRCD4938101G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26907-37-9
Molecular Formula C5 H6 N6 S2
Molecular Weight (g/mol) 214.2713
Synonym N,N'-Bis(1,3,4-thiadiazol-2-yl)methanediamine, 2,2'-(Methylenediimino)bis-1,3,4-thiadiazole, Bis-ADTA, Dikushuang, N,N'-Bis(1,3,4-thiadiazol-2-yl)methanediamine, N,N'-Methylenebis(2-amino-1,3,4-thiadiazole), NSC 143019
IUPAC Name N,N'-bis(1,3,4-thiadiazol-2-yl)methanediamine
SMILES C(Nc1nncs1)Nc2nncs2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H6N6S2/c1(6-4-10-8-2-12-4)7-5-11-9-3-13-5/h2-3H,1H2,(H,6,10)(H,7,11)
Formula Weight 214.0095
Chemical Name or Material N,N'-Di(1,3,4-thiadiazol-2-yl)methanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.