CAS RN 26907-37-9
CAS RN 26907-37-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N'-Di(1,3,4-thiadiazol-2-yl)methanediamine, TRC
CAS: 26907-37-9 Molecular Formula: C5 H6 N6 S2 Molecular Weight (g/mol): 214.2713 Synonym: N,N'-Bis(1,3,4-thiadiazol-2-yl)methanediamine,2,2'-(Methylenediimino)bis-1,3,4-thiadiazole,Bis-ADTA,Dikushuang,N,N'-Bis(1,3,4-thiadiazol-2-yl)methanediamine,N,N'-Methylenebis(2-amino-1,3,4-thiadiazole),NSC 143019 IUPAC Name: N,N'-bis(1,3,4-thiadiazol-2-yl)methanediamine SMILES: C(Nc1nncs1)Nc2nncs2