missing translation for 'onlineSavingsMsg'
Learn More

N-epsilon-Acetyl-L-lysine, 99%, MP Biomedicals™

Product Code. 11463041 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463041 1 g 1g
11473041 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463041 Supplier MP Biomedicals Supplier No. 0215023501
  • ONLINE EXCLUSIVE PRICE
    76.05 EUR
    72.45 EUR / 1g
    Save 3.60 EUR
    4% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 692-04-6
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00002639
InChI Key DTERQYGMUDWYAZ-ZETCQYMHSA-N
Synonym n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine
PubChem CID 92832
ChEBI CHEBI:17752
SMILES CC(=O)NCCCC[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 99%
IUPAC Name (2S)-2-amino-6-acetamidohexanoic acid
Quantity 1 g
Formula Weight 188.3
Physical Form Powder
Chemical Name or Material N-epsilon-acetyl-L-lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.