missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-aspartic acid 4-cyclohexyl ester, 95%, Thermo Scientific™

Product Code. 15439238
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439238 5 g 5g
15429238 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439238 Supplier Thermo Scientific Alfa Aesar Supplier No. H62758.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-D-aspartic acid 4-cyclohexyl ester is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112898-18-7
Molecular Formula C15H25NO6
Molecular Weight (g/mol) 315.37
MDL Number MFCD00060741
InChI Key NLPQIWFEEKQBBN-UHFFFAOYNA-N
Synonym boc-d-asp ochex-oh, boc-d-aspartic acid 4-cyslohexyl ester, boc-d-asp ochx-oh, r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, 2r-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid, boc-d-asp ochx-oh tlc, n-alpha-t-boc-d-aspartic beta-cyclohexyl ester, n-tert-butyloxycarbonyl-d-aspartic acid 4-cyslohexyl ester
PubChem CID 7016592
SMILES CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(O)=O

Specifications

Quantity 5 g
IUPAC Name (2R)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Formula Weight 315.36
Percent Purity 95%
Chemical Name or Material N-Boc-D-aspartic acid 4-cyclohexyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.