accessibility menu, dialog, popup

UserName

CH3CH3CH3OOHNOOOOH

CAS RN 112898-18-7

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-(cyclohexyloxy)-4-oxobutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-(cyclohexyloxy)-4-oxobutanoic acid
Molecular Weight (g/mol): 315.37
Molecular Formula: C15H25NO6
InChi Key: NLPQIWFEEKQBBN-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results