Learn More
N-Benzylaminoacetonitrile hydrochloride, 98%, Thermo Scientific™
Product Code. 11359415
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359415 5 g 5g
11369415 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11359415 Supplier Thermo Scientific Alfa Aesar Supplier No. L18566.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63086-36-2
Molecular Formula C9H11ClN2
Molecular Weight (g/mol) 182.651
MDL Number MFCD02094033
InChI Key OJUIRUMDLUMSTN-UHFFFAOYSA-N
Synonym 2-benzylamino acetonitrile hydrochloride, n-benzylaminoacetonitrile hydrochloride, n-benzylaminoacetonitrile, 2-benzylamino ;acetonitrile hydrochloride
PubChem CID 12571548
IUPAC Name 2-(benzylamino)acetonitrile;hydrochloride
SMILES C1=CC=C(C=C1)CNCC#N.Cl
Specifications
Melting Point 153°C to 160°C
Quantity 5 g
UN Number UN3439
Beilstein 3699545
Sensitivity Hygroscopic
Formula Weight 182.65
Percent Purity 98%
Chemical Name or Material N-Benzylaminoacetonitrile hydrochloride