accessibility menu, dialog, popup

UserName

HNNHCl

CAS RN 63086-36-2

IUPAC Name: 2-(benzylamino)acetonitrile hydrochloride
Synonyms: 2-(benzylamino)acetonitrile hydrochloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C9H11ClN2
InChi Key: OJUIRUMDLUMSTN-UHFFFAOYSA-N
SMILES: Cl.N#CCNCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results