missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-L-glutamine, 98+%, Thermo Scientific™

Product Code. 11332606
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332606 25 g 25g
11322606 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332606 Supplier Thermo Scientific Alfa Aesar Supplier No. L08604.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed in the synthesis and analgesic activity of human .beta.-endorphin.

Solubility
Soluble in DMSO and methanol. Soluble in 1 mmole in 2 ml DMF.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-85-7
Molecular Formula C10H17N2O5
Molecular Weight (g/mol) 245.26
MDL Number MFCD00065571
InChI Key VVNYDCGZZSTUBC-LURJTMIESA-M
Synonym boc-gln-oh, boc-l-glutamine, boc-glutamine, tert-butoxycarbonyl-l-glutamine, s-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, nalpha-tert-butoxycarbonyl-l-glutamine, n-tert-butoxycarbonyl-l-glutamine, boc-gln, nalpha-boc-l-glutamine, n2-tert-butoxycarbonyl-l-glutamine
PubChem CID 83687
IUPAC Name (2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCC(N)=O)C([O-])=O

Specifications

Melting Point 118°C to 120°C
Beilstein 2127805
Solubility Information Soluble in DMSO and methanol. Soluble in 1?mmole in 2 ml DMF.
Quantity 25 g
Formula Weight 246.26
Optical Rotation −18.5° (c=1, DMF)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-L-glutamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.