accessibility menu, dialog, popup

UserName

CAS RN 13726-85-7

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-carbamoylbutanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-4-carbamoylbutanoate
Molecular Weight (g/mol): 245.26
Molecular Formula: C10H17N2O5
InChi Key: VVNYDCGZZSTUBC-LURJTMIESA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CCC(N)=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results