missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-L-cysteine, >96%, MP Biomedicals™

Product Code. 11368399 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368399 25 g 25g
11378399 50 g 50g
11388399 100 g 100g
11398399 500 g 500g
11308409 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11368399 Supplier MP Biomedicals Supplier No. 0210009825
  • ONLINE EXCLUSIVE PRICE
    41.90 EUR
    39.90 EUR / 25g
    Save 2.00 EUR
    4% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.191
MDL Number MFCD00004880
InChI Key PWKSKIMOESPYIA-BYPYZUCNSA-N
Synonym n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PubChem CID 12035
ChEBI CHEBI:28939
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)NC(CS)C(=O)O

Specifications

Color White
Assay Percent Range >96%
Density 1.295g/cm3 (L)
Quantity 25 g
Formula Weight 163.2
Physical Form Crystalline Powder
Chemical Name or Material N-acetyl-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.