missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-β-D-mannosamine Hydrate, MP Biomedicals™

Product Code. 11338419 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338419 5 g 5g
11328419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338419 Supplier MP Biomedicals Supplier No. 0210010905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Acetyl-β-D-mannosamine Hydrate is an essential precursor of N-acetylneuraminic acid (NeuAc).

  • N-Acetyl-β-D-mannosamine hydrate is used as a substrate to identify, differentiate and characterize enzymes such as N-acetyl-glucosamine epimerase(s) and aldolase(s).
  • N-Acetyl-D-mannosamine is used for the synthesis of of N-acetylneuraminic acid.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Form Crystalline
Quantity 5 g
Product Type N-acetyl-β-D-Mannosamine
Purity or Quality Grade >98%

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.