missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Acetylphenyl)maleimide, 98%, Thermo Scientific™

Product Code. 11470843
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470843 25 g 25g
11450843 1 g 1g
11460843 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470843 Supplier Thermo Scientific Alfa Aesar Supplier No. A13282.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-(4-Acetylphenyl)maleimide is used as cross linking agent for reversible conjugation of thiol- containing compounds.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1082-85-5
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.21
MDL Number MFCD00014537
InChI Key ISGBKWSEHGAUNF-UHFFFAOYSA-N
Synonym 1-4-acetylphenyl-1h-pyrrole-2,5-dione, n-4-acetylphenyl maleimide, 1-4-acetylphenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-4-acetylphenyl, n-4-acetylphenyl maleinimide, 1-4-acetylphenyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-acetyl-4-2,5-dioxoazolinyl benzene, cyto8g9, p-maleimidoacetophenone, acetylphenylpyrroledione
PubChem CID 70660
IUPAC Name 1-(4-acetylphenyl)pyrrole-2,5-dione
SMILES CC(=O)C1=CC=C(C=C1)N1C(=O)C=CC1=O

Specifications

Melting Point 154°C to 158°C
Quantity 25 g
Beilstein 157371
Solubility Information Insoluble in water.
Formula Weight 215.21
Percent Purity 98%
Chemical Name or Material N-(4-Acetylphenyl)maleimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.