accessibility menu, dialog, popup

UserName

CAS RN 1082-85-5

H3CONOO

CAS RN 1082-85-5

IUPAC Name: 1-(4-acetylphenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-acetylphenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: ISGBKWSEHGAUNF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)N1C(=O)C=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results