missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-valine, 96%, Thermo Scientific™

Product Code. 15418685
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418685 250 mg 250mg
15408685 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418685 Supplier Thermo Scientific Alfa Aesar Supplier No. H33882.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 250714-85-3
Molecular Formula C12H14F3NO4S
Molecular Weight (g/mol) 325.30
MDL Number MFCD00173010
InChI Key KVNFXYSAEPLUHG-UHFFFAOYSA-N
Synonym 3-methyl-2-3-trifluoromethyl benzenesulfonamido butanoic acid, 3-trifluoromethyl phenyl sulfonyl valine, 3-methyl-2-3-trifluoromethyl phenyl sulfonyl amino butanoic acid, 3-methyl-2-3-trifluoromethyl phenyl-sulfonyl amino butanoic acid, valine,n-3-trifluoromethyl phenyl sulfonyl, 3-methyl-2-3-trifluoromethyl phenylsulfonamido butanoic acid, 3-methyl-2-3-trifluoromethyl phenyl sulfonylamino butanoic acid, n-3-trifluoromethyl phenyl sulfonyl valine
PubChem CID 2775542
IUPAC Name 3-methyl-2-[[3-(trifluoromethyl)phenyl]sulfonylamino]butanoic acid
SMILES CC(C)C(NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F)C(O)=O

Specifications

Melting Point 133°C to 134°C
Quantity 250 mg
Formula Weight 325.31
Percent Purity 96%
Chemical Name or Material N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.