accessibility menu, dialog, popup

UserName

CH3H3COOHHNSOOFFF

CAS RN 250714-85-3

IUPAC Name: 3-methyl-2-[3-(trifluoromethyl)benzenesulfonamido]butanoic acid
Synonyms: 3-methyl-2-[3-(trifluoromethyl)benzenesulfonamido]butanoic acid
Molecular Weight (g/mol): 325.3
Molecular Formula: C12H14F3NO4S
InChi Key: KVNFXYSAEPLUHG-UHFFFAOYNA-N
SMILES: CC(C)C(NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)