missing translation for 'onlineSavingsMsg'
Learn More

N-(2-(2,4,6-Trichlorophenoxy)ethyl)propan-1-amine, TRC

Product Code. 17971175
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971175 250 mg 250mg
17912365 50 mg 50mg
17951825 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971175 Supplier Toronto Research Chemicals Supplier No. TRCT895888250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67747-01-7
Molecular Formula C11 H14 Cl3 N O
Molecular Weight (g/mol) 282.59
Synonym N-[2-(2,4,6-Trichlorophenoxy)ethyl]-1-propanamine, N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]amine, N-[2-(2,4,6-Trichlorophenoxy)ethyl]propylamine
IUPAC Name N-[2-(2,4,6-trichlorophenoxy)ethyl]propan-1-amine
SMILES CCCNCCOc1c(Cl)cc(Cl)cc1Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H14Cl3NO/c1-2-3-15-4-5-16-11-9(13)6-8(12)7-10(11)14/h6-7,15H,2-5H2,1H3
Formula Weight 281.0141
Percent Purity >95%
Chemical Name or Material N-(2-(2,4,6-Trichlorophenoxy)ethyl)propan-1-amine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.