CAS RN 67747-01-7
CAS RN 67747-01-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-(2-(2,4,6-Trichlorophenoxy)ethyl)propan-1-amine, TRC
CAS: 67747-01-7 Molecular Formula: C11 H14 Cl3 N O Molecular Weight (g/mol): 282.59 Synonym: N-[2-(2,4,6-Trichlorophenoxy)ethyl]-1-propanamine,N-Propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]amine,N-[2-(2,4,6-Trichlorophenoxy)ethyl]propylamine IUPAC Name: N-[2-(2,4,6-trichlorophenoxy)ethyl]propan-1-amine SMILES: CCCNCCOc1c(Cl)cc(Cl)cc1Cl