missing translation for 'onlineSavingsMsg'
Learn More

MYT1L Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18214908 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214908 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214908 Supplier Novus Biologicals Supplier No. NBP281742AF532
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYT1L Polyclonal specifically detects MYT1L in Human/Mouse/Rat samples. It is validated for ELISA/Immunocytochemistry/Immunofluorescence/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYT1L
Applications ELISA, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias KIAA1106neural zinc finger transcription factor 1, myelin transcription factor 1-like, myelin transcription factor 1-like protein, MyT1L, MyT1-L, NZF1
Host Species Rabbit
Immunogen Rabbit polyclonal MYT1L antibody was raised against an 18 amino acid peptide near the carboxy terminus of human MYT1L. The immunogen is located within the last 50 amino acids of MYT1L. Amino Acid Squence: CTEMYTNQDRYQSPENKAL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23040
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.