Learn More
Novus Biologicals™ MUC20 293T Cell Transient Overexpression Lysate
Product Code. 18510023 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18510023 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18510023 Supplier Novus Biologicals™ Supplier No. H00200958T02
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Research Category Cell Biology, Cytoskeleton Markers, Signal Transduction
Target MUC20
Synonym FLJ14408, FLJ53153, KIAA1359mucin-20, MUC-20, mucin 20, cell surface associated, transmembrane mucin MUC20S
Gene Symbol MUC20