Learn More
R&D Systems™ Mpro 13b-K
Product Code. 18094892
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18094892 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18094892 Supplier R&D Systems™ Supplier No. 7228/5
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
A Tocris product, part of R&D Systems. Coronavirus Mpro inhibitor
Mpro 13b-K is a coronavirus Mpro inhibitor (IC50 values are 0.12, 0.58 and 0.9 μM for recombinant Mpro from SARS-CoV-2, MERS-CoV and SARS-CoV, respectively). Mpro 13b-K inhibits SARS-CoV-2 infection of human Calu3 cells (IC50 = 2.4 μM). The compound also inhibits SARS-CoV-2 replication in A549-ACE2-TMPRSS2, VeroE6 and Huh7 cell cultures (EC50 values are 0.84, 1.3 and 3.4 μM). Exhibits oral and inhalative bioavailability. Please note that this product was previously known as Mpro 13b.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material tert-Butyl (1-((S)-1-(((S)-4-(benzylamino)-3,4-dioxo-1-((S)-2-oxopyrrolidin-3-yl)butan-2-yl)amino)-3-cyclopropyl-1-oxopropan-2-yl)-2-oxo-1,2-dihydropyridin-3-yl)carbamate
CAS 2412965-59-2
Quantity 5 mg
Product Line Tocris
Target 3C and 3CL Protease Inhibitors
Molecular Formula C31H39N5O7
Purity 97%