Learn More
MP Biomedicals™ Freund's Incomplete Adjuvant
Product Code. 11434225 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434225 25 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11434225 Supplier MP Biomedicals™ Supplier No. 0855829
  • ONLINE EXCLUSIVE PRICE
    77.15 EUR
    73.50 EUR / 50mL
    Save 3.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Used with subsequent boosts to increase antibody titer following initial animal injection with Freund's Complete Adjuvant
Specifications
Content And Storage 2°C to 8°C
For Use With (Application) Used with subsequent boosts to increase antibody titer following initial animal injection with Freund's complete adjuvant, immunoassays, as boost injections
Formulation Paraffin oil 80 to 90%, Arlacel A 10 to 20%
Quantity 25 mL
Product Type Freund's Incomplete Adjuvant