Learn More
MP Biomedicals™ BCIP/NBT Stabilized Chromogen Solution, 100mL
Product Code. 11414346 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414346 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11414346 Supplier MP Biomedicals™ Supplier No. 08980621
  • ONLINE EXCLUSIVE PRICE
    89.30 EUR
    85.05 EUR / 100mL
    Save 4.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
For Use With (Application) Immunoassays
Quantity 100 mL
Content And Storage 0°C to 5°C
Product Type BCIP/NBT Stabilized Chromogen Solution