Learn More
Abcam Mouse small intestine tissue lysate - total protein, 100 ug
Product Code. 32143312 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32143312 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32143312 Supplier Abcam Supplier No. ab7939100ug
 Restricted Item
Estimated Shipment: 07-12-2026
Mouse small intestine cell/tissue lysate.
  • Species: Mouse
  • Tissue: small intestine
  • Form: Liquid
Specifications
For Use With (Application) Western Blot