Learn More
R&D Systems™ Mouse Granzyme C (NP_034501.2) VersaClone cDNA
Specifications
Specifications
| Gene | Granzyme C |
| Accession Number | NP_034501.2 |
| Gene ID (Entrez) | 14940 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |